Assay Details
| Gene Symbol: B3GALT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005601560.3 | XP_005601617.1 | 4444 | 67 | ||
| XM_014732518.2 | XP_014588004.1 | 6327 | 67 | |||
| XM_023623079.1 | XP_023478847.1 | 5970 | 67 | |||
| XM_023623082.1 | XP_023478850.1 | 546 | 67 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005601560.3, XM_014732518.2, XM_023623079.1, XM_023623082.1 transcripts.
Set Membership: |
|

B3GALT1
Genomic Map