Assay Details
Gene Symbol: TMEM35B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003364413.4 | XP_003364461.2 | 346 | 93 | ||
XM_023633760.1 | XP_023489528.1 | 1483 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
