Assay Details
| Gene Symbol: LOC100050294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023635700.1 | XP_023491468.1 | 1143 | 77 | ||
| XM_023635701.1 | XP_023491469.1 | 1065 | 77 | |||
| XM_023635702.1 | XP_023491470.1 | 1143 | 77 | |||
| XM_023635703.1 | XP_023491471.1 | 937 | 77 | |||
More Information
Set Membership: |
|

LOC100050294
Genomic Map