Assay Details
Gene Symbol: SPSB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494636.5 | XP_001494686.1 | 175 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001494636.5 transcripts.
Set Membership: |
![]() ![]() |
