Assay Details
Gene Symbol: NRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489822.5 | XP_001489872.1 | 420 | 90 | ||
XM_014734706.2 | XP_014590192.1 | 475 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
