Assay Details
| Gene Symbol: POLR1H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005603662.3 | XP_005603719.1 | 511 | 97 | ||
| XM_005603663.3 | XP_005603720.1 | 500 | 97 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

POLR1H| Gene Symbol: POLR1H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005603662.3 | XP_005603719.1 | 511 | 97 | ||
| XM_005603663.3 | XP_005603720.1 | 500 | 97 | |||
Set Membership: |
|