Assay Details
| Gene Symbol: C24H14orf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005605493.3 | XP_005605550.1 | 342 | 119 | ||
| XM_005605494.3 | XP_005605551.1 | 556 | 119 | |||
More Information
Set Membership: |
|

C24H14orf180
Genomic Map