Assay Details
| Gene Symbol: C25H9orf78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001499807.5 | XP_001499857.1 | 344 | 73 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C25H9orf78| Gene Symbol: C25H9orf78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001499807.5 | XP_001499857.1 | 344 | 73 | ||