Assay Details
Gene Symbol: LOC100069029 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498857.5 | XP_001498907.4 | 341 | 83 | ||
XM_023629287.1 | XP_023485055.1 | 341 | 83 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
