Assay Details
| Gene Symbol: C26H21orf58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014736497.2 | XP_014591983.1 | 373 | 80 | ||
| XM_023630343.1 | XP_023486111.1 | 375 | 80 | |||
| XM_023630344.1 | XP_023486112.1 | 375 | 80 | |||
More Information
Set Membership: | 
		   
					
						
							 | 
				

C26H21orf58
Genomic Map