Assay Details
| Gene Symbol: PRAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023630784.1 | XP_023486552.1 | 2646 | 110 | ||
| XM_023630785.1 | XP_023486553.1 | 2602 | 110 | |||
| XM_023630786.1 | XP_023486554.1 | 1995 | 110 | |||
More Information
Set Membership: |
|

PRAG1
Genomic Map