Assay Details
Gene Symbol: WASHC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014736782.2 | XP_014592268.1 | 2500 | 111 | ||
XM_023631814.1 | XP_023487582.1 | 2500 | 111 | |||
XM_023631815.1 | XP_023487583.1 | 2392 | 111 | |||
XM_023631816.1 | XP_023487584.1 | 2500 | 111 | |||
XM_023631817.1 | XP_023487585.1 | 1979 | 111 |

More Information
Set Membership: |
![]() ![]() |
