Assay Details
Gene Symbol: LOC100069464 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005606589.3 | XP_005606646.1 | 422 | 82 | ||
XM_005606590.3 | XP_005606647.1 | 306 | 82 | |||
XM_005606592.3 | XP_005606649.1 | 297 | 82 | |||
XM_023631869.1 | XP_023487637.1 | 225 | 82 | |||
XM_023631870.1 | XP_023487638.1 | 413 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005606589.3, XM_005606590.3, XM_005606592.3, XM_023631869.1, XM_023631870.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
