Assay Details
| Gene Symbol: LGALS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001501032.5 | XP_001501082.3 | 384 | 82 | ||
| GenBank mRNA | KY264050.1 | - | 2 - 3 | 261 | 82 | |
More Information
Set Membership: |
|

LGALS1
Genomic Map