Assay Details
| Gene Symbol: LOC100068023 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001497923.5 | XP_001497973.2 | 1429 | 109 | ||
| XM_005608398.3 | XP_005608455.1 | 1470 | 109 | |||
| XM_005608399.3 | XP_005608456.1 | 1386 | 109 | |||
| XM_023637263.1 | XP_023493031.1 | 1138 | 109 | |||
| XM_023637264.1 | XP_023493032.1 | 1409 | 109 | |||
| XM_023637265.1 | XP_023493033.1 | 1233 | 109 | |||
| XM_023637266.1 | XP_023493034.1 | 1266 | 109 | |||
| XM_023637267.1 | XP_023493035.1 | 999 | 109 | |||
| GenBank mRNA | JL625812.1 | - | 4 - 5 | 412 | 109 | |
More Information
Set Membership: |
|

LOC100068023
Genomic Map