Assay Details
| Gene Symbol: TMEM170A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003364637.4 | XP_003364685.2 | 653 | 63 | ||
| XM_023637345.1 | XP_023493113.1 | 1096 | 63 | |||
| XM_023637347.1 | XP_023493115.1 | 299 | 63 | |||
More Information
Set Membership: |
|

TMEM170A
Genomic Map