Assay Details
| Gene Symbol: MEAK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005608465.3 | XP_005608522.1 | 116 | 78 | ||
| XM_023637420.1 | XP_023493188.1 | 115 | 78 | |||
| XM_023637421.1 | XP_023493189.1 | 115 | 78 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005608465.3, XM_023637420.1, XM_023637421.1 transcripts.
Set Membership: |
|

MEAK7
Genomic Map