Assay Details
| Gene Symbol: MEAK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005608465.3 | XP_005608522.1 | 568 | 73 | ||
| XM_005608467.3 | XP_005608524.1 | 1220 | 73 | |||
| XM_014738373.2 | XP_014593859.1 | 1234 | 73 | |||
| XM_023637419.1 | XP_023493187.1 | 412 | 73 | |||
| XM_023637420.1 | XP_023493188.1 | 3031 | 73 | |||
| XM_023637421.1 | XP_023493189.1 | 1171 | 73 | |||
| XM_023637422.1 | XP_023493190.1 | 492 | 73 | |||

MEAK7
Genomic Map