Assay Details
Gene Symbol: PWWP3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014741248.2 | XP_014596734.1 | 1170 | 97 | ||
XM_023646292.1 | XP_023502060.1 | 1245 | 97 | |||
XM_023646293.1 | XP_023502061.1 | 1343 | 97 | |||
XM_023646294.1 | XP_023502062.1 | 1422 | 97 | |||
XM_023646295.1 | XP_023502063.1 | 1497 | 97 | |||
XM_023646296.1 | XP_023502064.1 | 1244 | 97 | |||
XM_023646297.1 | XP_023502065.1 | 1745 | 97 | |||
XM_023646298.1 | XP_023502066.1 | 1874 | 97 | |||
XM_023646299.1 | XP_023502067.1 | 1110 | 97 | |||
XM_023646300.1 | XP_023502068.1 | 1121 | 97 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
