Assay Details
Gene Symbol: LOC100630489 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005608718.3 | XP_005608775.1 | 234 | 56 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
