Assay Details
Gene Symbol: NSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014738599.2 | XP_014594085.1 | 4455 | 78 | ||
XM_023638510.1 | XP_023494278.1 | 3114 | 78 | |||
XM_023638511.1 | XP_023494279.1 | 3036 | 78 |
