Assay Details
Gene Symbol: LMOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495117.5 | XP_001495167.1 | 1894 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
