Assay Details
| Gene Symbol: LOC102147893 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014738131.2 | XP_014593617.1 | 2846 | 87 | ||
| XM_023632612.1 | XP_023488380.1 | 1920 | 87 | |||
More Information
Set Membership: |
|

LOC102147893
Genomic Map