Assay Details
Gene Symbol: LOC100064842 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005609110.3 | XP_005609167.1 | 237 | 101 | ||
XM_023638974.1 | XP_023494742.1 | 268 | 101 | |||
XM_023638975.1 | XP_023494743.1 | 243 | 101 | |||
XM_023638977.1 | XP_023494745.1 | 207 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005609110.3, XM_023638977.1 transcripts.
Set Membership: |
![]() ![]() |
