Assay Details
| Gene Symbol: CRB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001916887.5 | XP_001916922.2 | 975 | 91 | ||
| XM_023644582.1 | XP_023500350.1 | 991 | 91 | |||
| XM_023644583.1 | XP_023500351.1 | 1085 | 91 | |||
| XM_023644584.1 | XP_023500352.1 | 1060 | 91 | |||
| XM_023644585.1 | XP_023500353.1 | 1069 | 91 | |||
More Information
Set Membership: |
|

CRB3
Genomic Map