Assay Details
Gene Symbol: PET100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005611497.3 | XP_005611554.1 | 145 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005611497.3 transcripts.
Set Membership: |
![]() ![]() |
