Assay Details
| Gene Symbol: C7H11orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001499508.4 | XP_001499558.2 | 1283 | 107 | ||
| XM_023644747.1 | XP_023500515.1 | 1393 | 107 | |||
| XM_023644748.1 | XP_023500516.1 | 1397 | 107 | |||
| XM_023644749.1 | XP_023500517.1 | 1395 | 107 | |||
More Information
Set Membership: |
|

C7H11orf65
Genomic Map