Assay Details
Gene Symbol: C7H11orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001916879.4 | XP_001916914.1 | 165 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001916879.4 transcripts.
Set Membership: |
![]() ![]() |
