Assay Details
Gene Symbol: C7H11orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001916879.4 | XP_001916914.1 | 261 | 78 | ||
XM_005611559.3 | XP_005611616.1 | 180 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
