Assay Details
Gene Symbol: NXPE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005611582.3 | XP_005611639.1 | 636 | 69 | ||
XM_005611586.3 | XP_005611643.1 | 636 | 69 | |||
XM_014741370.2 | XP_014596856.1 | 624 | 69 | |||
XM_023644853.1 | XP_023500621.1 | 405 | 69 |

More Information
Set Membership: |
![]() ![]() |
