Assay Details
Gene Symbol: NACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504876.4 | XP_001504926.1 | 1470 | 89 | ||
XM_005611844.3 | XP_005611901.1 | 1472 | 89 | |||
XM_023645276.1 | XP_023501044.1 | 1501 | 89 | |||
XM_023645277.1 | XP_023501045.1 | 1397 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
