Assay Details
| Gene Symbol: CRPPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023639316.1 | XP_023495084.1 | 1095 | 88 | ||
| XM_023639317.1 | XP_023495085.1 | 1095 | 88 | |||
| XM_023639319.1 | XP_023495087.1 | 1095 | 88 | |||
| XM_023639320.1 | XP_023495088.1 | 1095 | 88 | |||
More Information
Set Membership: |
|

CRPPA
Genomic Map