Assay Details
Gene Symbol: SPON1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023646077.1 | XP_023501845.1 | 1458 | 92 | ||
XM_023646078.1 | XP_023501846.1 | 912 | 92 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
