Assay Details
Gene Symbol: NCR3LG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005612198.3 | XP_005612255.2 | 1143 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
