Assay Details
Gene Symbol: LOC100072189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023646186.1 | XP_023501954.1 | 461 | 88 | ||
XM_023646187.1 | XP_023501955.1 | 465 | 88 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100072189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023646186.1 | XP_023501954.1 | 461 | 88 | ||
XM_023646187.1 | XP_023501955.1 | 465 | 88 |