Assay Details
Gene Symbol: CCDC74B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023646670.1 | XP_023502438.1 | 812 | 85 | ||
XM_023646671.1 | XP_023502439.1 | 802 | 85 | |||
XM_023646672.1 | XP_023502440.1 | 769 | 85 | |||
XM_023646673.1 | XP_023502441.1 | 758 | 85 | |||
XM_023646674.1 | XP_023502442.1 | 811 | 85 | |||
XM_023646675.1 | XP_023502443.1 | 801 | 85 | |||
XM_023646676.1 | XP_023502444.1 | 696 | 85 | |||
XM_023646677.1 | XP_023502445.1 | 685 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
