Assay Details
| Gene Symbol: GATC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001489039.6 | XP_001489089.1 | 297 | 62 | ||
| XM_014742183.2 | XP_014597669.1 | 272 | 62 | |||
| GenBank mRNA | JL628420.1 | - | 1 - 2 | 40 | 62 | |
More Information
Set Membership: |
|

GATC
Genomic Map