Assay Details
Gene Symbol: LOC100055035 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023647059.1 | XP_023502827.1 | 1371 | 69 | ||
XM_023647060.1 | XP_023502828.1 | 1371 | 69 | |||
XM_023647061.1 | XP_023502829.1 | 1365 | 69 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
