Assay Details
| Gene Symbol: C8H18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498367.5 | XP_001498417.2 | 1171 | 91 | ||
| XM_003365544.4 | XP_003365592.1 | 1359 | 91 | |||
| XM_005612874.3 | XP_005612931.1 | 1196 | 91 | |||
| GenBank mRNA | JL638444.1 | - | 1 - 2 | 122 | 91 | |
More Information
Set Membership: |
|

C8H18orf25
Genomic Map