Assay Details
Gene Symbol: ZNF516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023647999.1 | XP_023503767.1 | 3724 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
