Assay Details
| Gene Symbol: ELOC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001493018.6 | XP_001493068.2 | 360 | 106 | ||
| XM_003365580.4 | XP_003365628.1 | 342 | 106 | |||
| XM_005613051.3 | XP_005613108.1 | 223 | 106 | |||
More Information
Set Membership: |
|

ELOC
Genomic Map