Assay Details
Gene Symbol: LOC100066505 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496755.4 | XP_001496805.1 | 150 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001496755.4 transcripts.
Set Membership: |
![]() ![]() |
