Assay Details
Gene Symbol: LY6E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496550.5 | XP_001496600.5 | 380 | 69 | ||
XM_005613333.3 | XP_005613390.2 | 368 | 69 | |||
XM_023649068.1 | XP_023504836.1 | 437 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
