Assay Details
Gene Symbol: ZNF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005613372.3 | XP_005613429.1 | 603 | 74 | ||
XM_005613373.3 | XP_005613430.1 | 691 | 74 | |||
XM_005613376.3 | XP_005613433.1 | 53 | 74 | |||
XM_023649314.1 | XP_023505082.1 | 443 | 74 | |||
XM_023649315.1 | XP_023505083.1 | 397 | 74 | |||
XM_023649316.1 | XP_023505084.1 | 960 | 74 | |||
XM_023649317.1 | XP_023505085.1 | 956 | 74 | |||
XM_023649318.1 | XP_023505086.1 | 598 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005613376.3, XM_023649318.1 transcripts.
Set Membership: |
![]() ![]() |
