Assay Details
Gene Symbol: LOC100053137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023646633.1 | XP_023502401.1 | 3104 | 80 | ||
XM_023646634.1 | XP_023502402.1 | 2990 | 80 | |||
XM_023646635.1 | XP_023502403.1 | 3003 | 80 | |||
XM_023646636.1 | XP_023502404.1 | 2949 | 80 | |||
XM_023646637.1 | XP_023502405.1 | 3098 | 80 | |||
XM_023646638.1 | XP_023502406.1 | 3071 | 80 | |||
XM_023646639.1 | XP_023502407.1 | 3044 | 80 | |||
XM_023646640.1 | XP_023502408.1 | 2996 | 80 | |||
XM_023646641.1 | XP_023502409.1 | 3104 | 80 | |||
XM_023646642.1 | XP_023502410.1 | 2915 | 80 | |||
XM_023646643.1 | XP_023502411.1 | 2356 | 80 | |||
XM_023646644.1 | XP_023502412.1 | 2168 | 80 | |||
XM_023646645.1 | XP_023502413.1 | 3363 | 80 | |||
XM_023646646.1 | XP_023502414.1 | 2018 | 80 | |||
XM_023646647.1 | XP_023502415.1 | 3527 | 80 | |||
XM_023646648.1 | XP_023502416.1 | 1928 | 80 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
