Assay Details
Gene Symbol: LOC100053137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023646633.1 | XP_023502401.1 | 3227 | 61 | ||
XM_023646634.1 | XP_023502402.1 | 3113 | 61 | |||
XM_023646635.1 | XP_023502403.1 | 3126 | 61 | |||
XM_023646636.1 | XP_023502404.1 | 3072 | 61 | |||
XM_023646637.1 | XP_023502405.1 | 3221 | 61 | |||
XM_023646638.1 | XP_023502406.1 | 3194 | 61 | |||
XM_023646639.1 | XP_023502407.1 | 3167 | 61 | |||
XM_023646640.1 | XP_023502408.1 | 3119 | 61 | |||
XM_023646641.1 | XP_023502409.1 | 3227 | 61 | |||
XM_023646642.1 | XP_023502410.1 | 3038 | 61 | |||
XM_023646643.1 | XP_023502411.1 | 2479 | 61 | |||
XM_023646644.1 | XP_023502412.1 | 2291 | 61 | |||
XM_023646645.1 | XP_023502413.1 | 3486 | 61 | |||
XM_023646646.1 | XP_023502414.1 | 2141 | 61 | |||
XM_023646647.1 | XP_023502415.1 | 3650 | 61 | |||
XM_023646648.1 | XP_023502416.1 | 2051 | 61 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
