Assay Details
Gene Symbol: LOC100053137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023646633.1 | XP_023502401.1 | 3321 | 64 | ||
XM_023646634.1 | XP_023502402.1 | 3207 | 64 | |||
XM_023646635.1 | XP_023502403.1 | 3220 | 64 | |||
XM_023646636.1 | XP_023502404.1 | 3166 | 64 | |||
XM_023646637.1 | XP_023502405.1 | 3315 | 64 | |||
XM_023646638.1 | XP_023502406.1 | 3288 | 64 | |||
XM_023646639.1 | XP_023502407.1 | 3261 | 64 | |||
XM_023646640.1 | XP_023502408.1 | 3213 | 64 | |||
XM_023646641.1 | XP_023502409.1 | 3321 | 64 | |||
XM_023646642.1 | XP_023502410.1 | 3132 | 64 | |||
XM_023646643.1 | XP_023502411.1 | 2573 | 64 | |||
XM_023646644.1 | XP_023502412.1 | 2385 | 64 | |||
XM_023646645.1 | XP_023502413.1 | 3580 | 64 | |||
XM_023646646.1 | XP_023502414.1 | 2235 | 64 | |||
XM_023646647.1 | XP_023502415.1 | 3744 | 64 | |||
XM_023646648.1 | XP_023502416.1 | 2145 | 64 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
