Assay Details
| Gene Symbol: BRME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005613573.3 | XP_005613630.1 | 155 | 96 | ||
| XM_005613574.3 | XP_005613631.1 | 136 | 96 | |||
| XM_005613575.3 | XP_005613632.1 | 222 | 96 | |||
| XM_023645257.1 | XP_023501025.1 | 111 | 96 | |||
| XM_023645258.1 | XP_023501026.1 | 154 | 96 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005613573.3, XM_005613574.3, XM_005613575.3, XM_023645257.1, XM_023645258.1 transcripts.
Set Membership: |
|

BRME1
Genomic Map