Assay Details
Gene Symbol: PABIR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005614514.3 | XP_005614571.1 | 139 | 83 | ||
XM_005614515.3 | XP_005614572.1 | 139 | 83 | |||
XM_014729197.2 | XP_014584683.1 | 139 | 83 |
