Assay Details
| Gene Symbol: PABIR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005614514.3 | XP_005614571.1 | 507 | 63 | ||
| XM_005614515.3 | XP_005614572.1 | 507 | 63 | |||
| XM_005614516.3 | XP_005614573.1 | 446 | 63 | |||
| XM_014729197.2 | XP_014584683.1 | 507 | 63 | |||
| XM_014729199.2 | XP_014584685.1 | 503 | 63 | |||
| GenBank mRNA | JL636786.1 | - | 5 - 6 | 381 | 63 | |
More Information
Set Membership: |
|

PABIR3
Genomic Map